Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Oxytetracycline, 'can Be Used as Secondarystandard'

Catalog No. AC257705000
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC257705000 500 mg Glass Bottle
AC257700025 2.5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC257705000 Supplier Thermo Scientific Chemicals Supplier No. 257705000
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-57-2
Molecular Formula C22H26N2Na2O11
Molecular Weight (g/mol) 540.43
MDL Number MFCD00003700
InChI Key OLRBUPVCLOPDOT-GETZKYKJSA-M
PubChem CID 54675779
IUPAC Name (4S,4aR,5S,5aR,6S,12aR)-4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide
SMILES O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O

Specifications

Type Oxytetracycline
Physical Form Crystalline Powder
Melting Point 183.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation − 223.00
Color Beige to Yellow
Quantity 500 mg
Solubility Information Solubility in water: 0.2g/L. Other solubilities: soluble in acids and alkalies, 25 mg/mL methanol, slightly soluble in acetone and chloroform, practically insoluble in ether
Formula Weight 460.43
Percent Purity ≥80 wt% (as is, calculated) (HPLC)
Chemical Name or Material Oxytetracycline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.