CAS RN 79-63-0
IUPAC Name:
(1R,3aR,5aR,7S,9aS,11aR)-3a,6,6,9a,11a-pentamethyl-1-[(2R)-6-methylhept-5-en-2-yl]-1H,2H,3H,3aH,4H,5H,5aH,6H,7H,8H,9H,9aH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Synonyms:
lanosterol (3β)-lanosta-8,24-dien-3-ol
Molecular Weight (g/mol):
426.73
Molecular Formula:
C30H50O
InChi Key:
CAHGCLMLTWQZNJ-BQNIITSRSA-N
SMILES:
C[C@H](CCC=C(C)C)[C@H]1CC[C@@]2(C)C3=C(CC[C@]12C)[C@@]1(C)CC[C@H](O)C(C)(C)[C@@H]1CC3
1
–
1
of
1
results
1
Thermo Scientific Chemicals Lanosterol, 50%
CAS: 79-63-0 Molecular Formula: C30H50O