accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79-95-8

IUPAC Name: 2,6-dichloro-4-[2-(3,5-dichloro-4-hydroxyphenyl)propan-2-yl]phenol
Synonyms: tetrachlorobisphenol A tetrachlordian tetrachlorodian
Molecular Weight (g/mol): 366.06
Molecular Formula: C15H12Cl4O2
InChi Key: KYPYTERUKNKOLP-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC(Cl)=C(O)C(Cl)=C1)C1=CC(Cl)=C(O)C(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results