Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachlorobisphenol A, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015566 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5566 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-5566 Supplier MilliporeSigma Supelco Supplier No. 7308110MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

CAS 79-95-8
Linear Formula (CH3)2 C[C6H2(Cl)2 OH]2
MDL Number MFCD00002178
Melting Point 134°C to 136°C
Synonym 4,4 ′-Isopropylidenebis(2,6-dichlorophenol); 2,2 ′,6,6 ′-Tetrachloro-4,4 ′-isopropylidenediphenol
Type Analytical Standard
Quantity 10 mg
Formula Weight 366.07
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.