accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79-97-0

IUPAC Name: 4-[2-(4-hydroxy-3-methylphenyl)propan-2-yl]-2-methylphenol
Synonyms: bisphenol C 3,3'-dimethyldian 3,3'-dimethylbisphenol A nonox dcp dicresylolpropane
Molecular Weight (g/mol): 256.35
Molecular Formula: C17H20O2
InChi Key: YMTYZTXUZLQUSF-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1O)C(C)(C)C1=CC=C(O)C(C)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)