accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79069-50-4

CH3(S)ONHOOH3CH3CCH3

CAS RN 79069-50-4

IUPAC Name: tert-butyl N-[(2S)-1-oxopropan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S)-1-oxopropan-2-yl]carbamate
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: OEQRZPWMXXJEKU-LURJTMIESA-N
SMILES: C[C@H](NC(=O)OC(C)(C)C)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results