Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-L-alaninal, 98%

Catalog No. AC438470010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438470010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC438470010 Supplier Thermo Scientific Chemicals Supplier No. 438470010
Add to Cart

CAS: 79069-50-4 | C8H15NO3 | 173.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79069-50-4
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00143786
InChI Key OEQRZPWMXXJEKU-LURJTMIESA-N
Synonym boc-l-alaninal, boc-ala-aldehyde, s-tert-butyl 1-oxopropan-2-yl carbamate, boc-l-alanine aldehyde, tert-butyl n-2s-1-oxopropan-2-yl carbamate, s-tert-butyl 1-oxopropan-2-ylcarbamate, chembl94727, s-1-methyl-2-oxo-ethyl-carbamic acid tert-butyl ester, carbamic acid, 1s-1-methyl-2-oxoethyl-, 1,1-dimethylethyl ester, 1-methyl-2-oxo-ethyl-carbamic acid tert-butyl ester
PubChem CID 7015728
IUPAC Name tert-butyl N-[(2S)-1-oxopropan-2-yl]carbamate
SMILES C[C@H](NC(=O)OC(C)(C)C)C=O

Specifications

Melting Point 86.4°C to 87.6°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Specific Rotation -30.6
Quantity 1 g
Formula Weight 173.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material BOC-L-Alaninal
Show More Show Less
Hazard Statement GHS H Statement:
Precautionary Statement GHS P Statement:
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.