accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79261-58-8

IUPAC Name: (3S)-2-[(benzyloxy)carbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Synonyms: (3S)-2-[(benzyloxy)carbonyl]-3,4-dihydro-1H-isoquinoline-3-carboxylate
Molecular Weight (g/mol): 310.33
Molecular Formula: C18H16NO4
InChi Key: YWVQGUBCAUFBCP-INIZCTEOSA-M
SMILES: [O-]C(=O)[C@@H]1CC2=CC=CC=C2CN1C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results