accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3NS

CAS RN 79265-36-4

IUPAC Name: 5-(trimethylsilyl)-1,3-thiazole
Synonyms: 5-(trimethylsilyl)-1,3-thiazole
Molecular Weight (g/mol): 157.31
Molecular Formula: C6H11NSSi
InChi Key: BFCGWRVEKWHDGG-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CN=CS1
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)