Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Trimethylsilylthiazole 98.0+%, TCI America™
SDP

Catalog No. T15231ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15231ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T15231ML Supplier TCI America Supplier No. T15231ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79265-36-4
Molecular Formula C6H11NSSi
Molecular Weight (g/mol) 157.31
MDL Number MFCD00191506
InChI Key BFCGWRVEKWHDGG-UHFFFAOYSA-N
PubChem CID 2760867
IUPAC Name 5-(trimethylsilyl)-1,3-thiazole
SMILES C[Si](C)(C)C1=CN=CS1

Specifications

Color Yellow
Boiling Point 65°C
Quantity 1 mL
Formula Weight 157.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Trimethylsilylthiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.