accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 79534-91-1

IUPAC Name: 7,14,25,32-tetraazaundecacyclo[21.13.2.2²,⁵.0³,¹⁹.0⁴,¹⁶.0⁶,¹⁴.0⁸,¹³.0²⁰,³⁷.0²⁴,³².0²⁶,³¹.0³⁴,³⁸]tetraconta-1(36),2(40),3,5(39),6,8,10,12,16,18,20,22,24,26,28,30,34,37-octadecaene-15,33-dione; 7,14,25,32-tetraazaundecacyclo[21.13.2.2²,⁵.0³,¹⁹.0⁴,¹⁶.0⁶,¹⁴.0⁸,¹³.0²⁰,³⁷.0²⁵,³³.0²⁶,³¹.0³⁴,³⁸]tetraconta-1(36),2(40),3,5(39),6,8,10,12,16,18,20,22,26,28,30,32,34,37-octadecaene-15,24-dione
Synonyms: 7,14,25,32-tetraazaundecacyclo[21.13.2.2²,⁵.0³,¹⁹.0⁴,¹⁶.0⁶,¹⁴.0⁸,¹³.0²⁰,³⁷.0²⁴,³².0²⁶,³¹.0³⁴,³⁸]tetraconta-1(36),2(40),3,5(39),6,8,10,12,16,18,20,22,24,26,28,30,34,37-octadecaene-15,33-dione; 7,14,25,32-tetraazaundecacyclo[21.13.2.2²,⁵.0³,¹⁹.0⁴,¹⁶.0⁶,¹⁴.0⁸,¹³.0²⁰,³⁷.0²⁵,³³.0²⁶,³¹.0³⁴,³⁸]tetraconta-1(36),2(40),3,5(39),6,8,10,12,16,18,20,22,26,28,30,32,34,37-octadecaene-15,24-dione
Molecular Weight (g/mol): 1073.1
Molecular Formula: C72H32N8O4
InChi Key: XQNMSKCVXVXEJT-UHFFFAOYSA-N
SMILES: O=C1N2C3=CC=CC=C3N=C2C2=CC=C3C4=CC=C5C(=O)N6C7=CC=CC=C7N=C6C6=CC=C(C7=CC=C1C2=C37)C4=C56.O=C1N2C3=CC=CC=C3N=C2C2=CC=C3C4=CC=C5C6=NC7=CC=CC=C7N6C(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results