Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTCBI (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. P2119200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2119200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2119200MG Supplier TCI America Supplier No. P2119200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79534-91-1
Molecular Formula C72H32N8O4
Molecular Weight (g/mol) 1073.1
InChI Key XQNMSKCVXVXEJT-UHFFFAOYSA-N
Synonym 3,4,9,10-Perylenetetracarboxylic Bisbenzimidazole, Anthra[9,1,2-c,d,e:10,5,6-c′C,d′C,e′C][bis[benzimidazolo[2,1-a]isoquinoline]]-10,21-dione
PubChem CID 91972167
SMILES C1=CC=C2C(=C1)N=C3N2C(=O)C4=CC=C5C6=CC=C7C8=C(C=CC(=C68)C9=C5C4=C3C=C9)C1=NC2=CC=CC=C2N1C7=O.C1=CC=C2C(=C1)N=C3N2C(=O)C4=CC=C5C6=C7C(=CC=C8C7=C(C=C6)C9=NC1=CC=CC=C1N9C8=O)C1=C5C4=C3C=C1

Specifications

Color Black
Quantity 200 mg
Formula Weight 536.55
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material PTCBI (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.