CAS RN 79580-28-2
CAS RN 79580-28-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Brevetoxin PbTx-2, Ptychodiscus brevis
Lipid-soluble polyether neurotoxin that binds to Na+ channels on nerves and muscles causing an excessive influx of Na+ across membranes.