accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClNHNClFFFNOOFFF

CAS RN 79622-59-6

IUPAC Name: 3-chloro-N-[3-chloro-2,6-dinitro-4-(trifluoromethyl)phenyl]-5-(trifluoromethyl)pyridin-2-amine
Synonyms: fluazinam
Molecular Weight (g/mol): 465.09
Molecular Formula: C13H4Cl2F6N4O4
InChi Key: UZCGKGPEKUCDTF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=C(Cl)C(=C1NC1=NC=C(C=C1Cl)C(F)(F)F)[N+]([O-])=O)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results