Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluazinam 98.0+%, TCI America™
SDP

Catalog No. F08691G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08691G Supplier TCI America Supplier No. F08691G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79622-59-6
Molecular Formula C13H4Cl2F6N4O4
Molecular Weight (g/mol) 465.09
MDL Number MFCD00214168
InChI Key UZCGKGPEKUCDTF-UHFFFAOYSA-N
Synonym fluazinam, frowncide, shirlan flow, altima, sekoya, mapro, shirlan zeneca, fluazinam iso, unii-0p91pck33q, 3-chloro-n-3-chloro-2,6-dinitro-4-trifluoromethyl phenyl-5-trifluoromethyl pyridin-2-amine
PubChem CID 91731
ChEBI CHEBI:81843
IUPAC Name 3-chloro-N-[3-chloro-2,6-dinitro-4-(trifluoromethyl)phenyl]-5-(trifluoromethyl)pyridin-2-amine
SMILES [O-][N+](=O)C1=CC(=C(Cl)C(=C1NC1=NC=C(C=C1Cl)C(F)(F)F)[N+]([O-])=O)C(F)(F)F

Specifications

Melting Point 118°C
Color Yellow
Quantity 1 g
UN Number 2811
Formula Weight 465.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fluazinam
RTECSNumber UR8085000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.