accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrOHO

CAS RN 79669-49-1

IUPAC Name: 5-bromo-2-methylbenzoic acid
Synonyms: 5-bromo-2-methylbenzoic acid
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: SEENCYZQHCUTSB-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)C=C1C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)