Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH6051606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6051606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6051606 Supplier Thermo Scientific Chemicals Supplier No. H6051606
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 79669-49-1 | Molecular Formula: C8H7BrO2 | Molecular Weight: 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79669-49-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00267350
InChI Key SEENCYZQHCUTSB-UHFFFAOYSA-N
Synonym 2-methyl-5-bromobenzoic acid, 5-bromo-o-toluic acid, 5-bromo-2-methyl-benzoic acid, 5-bromo-2-methyl benzoic acid, synquest 2721-9-x8, benzoic acid, 5-bromo-2-methyl, pubchem4728, acmc-1bhwo, 3-bromo-6-methylbenzoic acid, ksc497q7f
PubChem CID 346004
IUPAC Name 5-bromo-2-methylbenzoic acid
SMILES CC1=CC=C(Br)C=C1C(O)=O

Specifications

Melting Point 167°C to 171°C
Quantity 5 g
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methylbenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.