CAS RN 796967-10-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ VEGFR2/Flt3/c-Kit Inhibitor
The VEGFR2/Flt3/c-Kit Inhibitor, also referenced under CAS 796967-10-7, controls the biological activity of VEGFR2/Flt3/c-Kit. This small molecule/inhibitor is primarily used for Activators/Inducers applications.