Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ VEGFR2/Flt3/c-Kit Inhibitor

Catalog No. 6765005MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-650-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-650-05MG Supplier MilliporeSigma™ Supplier No. 6765005MG
Add to Cart
Add to Cart

The VEGFR2/Flt3/c-Kit Inhibitor, also referenced under CAS 796967-10-7, controls the biological activity of VEGFR2/Flt3/c-Kit. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 97% by HPLC
CAS 796967-10-7
Molecular Formula C21H19N5O
Formula Weight 357.4g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.