CAS RN 79791-38-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dodecylbenzenesulfonyl Azide (soft type) (mixture), TCI America™
CAS: 79791-38-1 Molecular Formula: C18H29N3O2S MDL Number: MFCD00274161