Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylbenzenesulfonyl Azide (soft type) (mixture), TCI America™
SDP

Catalog No. D258025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D258025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D258025G Supplier TCI America Supplier No. D258025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79791-38-1
Molecular Formula C18H29N3O2S
MDL Number MFCD00274161

Specifications

Quantity 25 g
Formula Weight 351.51
Physical Form Liquid
Chemical Name or Material Dodecylbenzenesulfonyl Azide (soft type) (mixture)
EINECSNumber (3)-4380
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.