CAS RN 79886-55-8
IUPAC Name:
2,5-dioxopyrrolidin-1-yl 4-[4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)phenyl]butanoate
Synonyms:
2,5-dioxopyrrolidin-1-yl 4-[4-(2,5-dioxopyrrol-1-yl)phenyl]butanoate
Molecular Weight (g/mol):
356.33
Molecular Formula:
C18H16N2O6
InChi Key:
PMJWDPGOWBRILU-UHFFFAOYSA-N
SMILES:
O=C(CCCC1=CC=C(C=C1)N1C(=O)C=CC1=O)ON1C(=O)CCC1=O
1
–
1
of
1
results
1
N-Succinimidyl 4-(4-Maleimidophenyl)butyrate 97.0+%, TCI America™
CAS: 79886-55-8 Molecular Formula: C18H16N2O6 Molecular Weight (g/mol): 356.334 MDL Number: MFCD00054961 InChI Key: PMJWDPGOWBRILU-UHFFFAOYSA-N Synonym: 4-(4-Maleimidophenyl)butyric Acid N-Succinimidyl Ester, SMPB PubChem CID: 100681 IUPAC Name: (2,5-dioxopyrrolidin-1-yl) 4-[4-(2,5-dioxopyrrol-1-yl)phenyl]butanoate SMILES: C1CC(=O)N(C1=O)OC(=O)CCCC2=CC=C(C=C2)N3C(=O)C=CC3=O