Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 4-(4-Maleimidophenyl)butyrate 97.0+%, TCI America™
SDP

Catalog No. S086110MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S086110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S086110MG Supplier TCI America Supplier No. S086110MG

Chemical Identifiers

CAS 79886-55-8
Molecular Formula C18H16N2O6
Molecular Weight (g/mol) 356.334
MDL Number MFCD00054961
InChI Key PMJWDPGOWBRILU-UHFFFAOYSA-N
Synonym 4-(4-Maleimidophenyl)butyric Acid N-Succinimidyl Ester, SMPB
PubChem CID 100681
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-[4-(2,5-dioxopyrrol-1-yl)phenyl]butanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCCC2=CC=C(C=C2)N3C(=O)C=CC3=O

Specifications

Melting Point 131°C
Quantity 10 mg
Formula Weight 356.33
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 4-(4-Maleimidophenyl)butyrate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.