accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 799557-86-1

NNFFFBr

CAS RN 799557-86-1

IUPAC Name: 5-bromo-2-(trifluoromethyl)pyrimidine
Synonyms: 5-bromo-2-(trifluoromethyl)pyrimidine
Molecular Weight (g/mol): 226.98
Molecular Formula: C5H2BrF3N2
InChi Key: GIFDWXWNFKZVEI-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC=C(Br)C=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results