accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOClCl

CAS RN 80-06-8

IUPAC Name: 1,1-bis(4-chlorophenyl)ethan-1-ol
Synonyms: dimite milbex
Molecular Weight (g/mol): 267.15
Molecular Formula: C14H12Cl2O
InChi Key: URYAFVKLYSEINW-UHFFFAOYSA-N
SMILES: CC(O)(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)