Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dichloro-alpha-methylbenzhydrol, TCI America™
SDP

Catalog No. D269225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D269225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D269225G Supplier TCI America Supplier No. D269225G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 80-06-8
Molecular Formula C14H12Cl2O
Molecular Weight (g/mol) 267.15
MDL Number MFCD00000628
InChI Key URYAFVKLYSEINW-UHFFFAOYSA-N
Synonym chlorfenethol, 4,4'-dichloro-alpha-methylbenzhydrol, dimit, 1,1-bis 4-chlorophenyl ethanol, micasin, mikazene, quickron, qikron, dimite, bcpe
PubChem CID 6624
ChEBI CHEBI:82154
IUPAC Name 1,1-bis(4-chlorophenyl)ethan-1-ol
SMILES CC(O)(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 267.15
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dichloro-alpha-methylbenzhydrol
RTECSNumber DC7875000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.