CAS RN 80-11-5
IUPAC Name:
N,4-dimethyl-N'-oxobenzene-1-sulfonohydrazide
Synonyms:
diazald
Molecular Weight (g/mol):
214.24
Molecular Formula:
C8H10N2O3S
InChi Key:
FFKZOUIEAHOBHW-UHFFFAOYSA-N
SMILES:
CN(N=O)S(=O)(=O)C1=CC=C(C)C=C1
1
–
1
of
1
results
1
Sigma Aldrich Imidazolyltetrafluorobromoethane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.