accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSOO

CAS RN 80-18-2

IUPAC Name: methyl benzenesulfonate
Synonyms: methyl benzenesulfonate
Molecular Weight (g/mol): 172.2
Molecular Formula: C7H8O3S
InChi Key: CZXGXYBOQYQXQD-UHFFFAOYSA-N
SMILES: COS(=O)(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (8)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (2)
  • (8)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)