accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHSOOH2NNN

CAS RN 80-35-3

IUPAC Name: 4-amino-N-(6-methoxypyridazin-3-yl)benzene-1-sulfonamide
Synonyms: longin durox davosin sulfozona kynex retamid paramid sulphamethoxypyridazine 磺胺甲氧嗪 kinex
Molecular Weight (g/mol): 280.3
Molecular Formula: C11H12N4O3S
InChi Key: VLYWMPOKSSWJAL-UHFFFAOYSA-N
SMILES: COC1=CC=C(NS(=O)(=O)C2=CC=C(N)C=C2)N=N1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)