Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamethoxypyridazine, Thermo Scientific Chemicals

Catalog No. AC436470250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436470250 25 g Glass bottle
AC436470050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC436470250 Supplier Thermo Scientific Chemicals Supplier No. 436470250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 80-35-3 | Molecular Formula: C11H12N4O3S | Molecular Weight: 280.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-35-3
Molecular Formula C11H12N4O3S
Molecular Weight (g/mol) 280.3
InChI Key VLYWMPOKSSWJAL-UHFFFAOYSA-N
Synonym sulfamethoxypyridazine, sulphamethoxypyridazine, sulfapyridazine, sulfalex, longin, midicel, sulfametoxipiridazine, spofadazine, sulfdurazin, 3-sulfa-6-methoxypyridazine
PubChem CID 5330
ChEBI CHEBI:102516
IUPAC Name 4-amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide
SMILES COC1=NN=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Melting Point 182°C to 183°C
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 g
Merck Index 15,9051
Solubility Information Solubility in water: insoluble
Formula Weight 280.3
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Sulfamethoxypyridazine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-272-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.