accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80-49-9

IUPAC Name: 3-[(2-hydroxy-2-phenylacetyl)oxy]-8,8-dimethyl-8-azabicyclo[3.2.1]octan-8-ium bromide
Synonyms: methylhomatropine bromide metilbromuro de homatropina tropinium methobromide mandelate methylbromure d'homatropine 8-methylhomatropinium bromide methylhomatropinum bromatum omatropina metilbromuro homatropini methylbromidum
Molecular Weight (g/mol): 370.29
Molecular Formula: C17H24BrNO3
InChi Key: FUFVKLQESJNNAN-UHFFFAOYNA-M
SMILES: [Br-].C[N+]1(C)C2CCC1CC(C2)OC(=O)C(O)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results