Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Homatropine Methyl Bromide, TCI America™
SDP

Catalog No. H04485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04485G Supplier TCI America Supplier No. H04485G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 80-49-9
Molecular Formula C17H24BrNO3
Molecular Weight (g/mol) 370.287
MDL Number MFCD00050292
InChI Key FUFVKLQESJNNAN-BXSBZORQSA-M
Synonym N-Methylhomatropinium Bromide
PubChem CID 73416022
IUPAC Name [(1S,5R)-8,8-dimethyl-8-azoniabicyclo[3.2.1]octan-3-yl] (2R)-2-hydroxy-2-phenylacetate;bromide
SMILES C[N+]1(C2CCC1CC(C2)OC(=O)C(C3=CC=CC=C3)O)C.[Br-]

Specifications

Melting Point 196°C
Color White
Quantity 5 g
UN Number 1544
Formula Weight 370.29
Physical Form Crystalline Powder
Chemical Name or Material Homatropine Methyl Bromide
RTECSNumber MS9000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.