accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80-82-0

IUPAC Name: 2-nitrobenzene-1-sulfonic acid
Synonyms: 2-nitrobenzenesulfonic acid
Molecular Weight (g/mol): 203.17
Molecular Formula: C6H5NO5S
InChi Key: HWTDMFJYBAURQR-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results