accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OH(S)(S)(R)(R)(R)OH(S)H3C(S)(S)H3C

CAS RN 80-92-2

IUPAC Name: (1S,3aS,3bR,5aR,7R,9aS,9bS,11aS)-1-[(1S)-1-hydroxyethyl]-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-7-ol
Synonyms: pregnanediol
Molecular Weight (g/mol): 320.52
Molecular Formula: C21H36O2
InChi Key: YWYQTGBBEZQBGO-BERLURQNSA-N
SMILES: C[C@H](O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results