Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5β-Pregnane-3α,20α-diol 99%, Thermo Scientific™

Catalog No. AC215581000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC215581000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC215581000 Supplier Thermo Scientific Chemicals Supplier No. 215581000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-92-2
Molecular Formula C21H36O2
Molecular Weight (g/mol) 320.517
InChI Key YWYQTGBBEZQBGO-BERLURQNSA-N
Synonym pregnanediol, pregnandiol, unii-jr3jd1y22c, 5beta-pregnane-3alpha,20alpha-diol, jr3jd1y22c, pregnandiol jan, pregnane-3,20-diol, pregnane-3,20-diol, 3a,5b,20s, allopregnanediol, pregnane-3alpha,20alpha-diol
PubChem CID 219833
ChEBI CHEBI:8387
IUPAC Name (3R,5R,8R,9S,10S,13S,14S,17S)-17-[(1S)-1-hydroxyethyl]-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C1CCC2C1(CCC3C2CCC4C3(CCC(C4)O)C)C)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 238°C to 241°C
Color White
Infrared Spectrum Authentic
Quantity 100 mg
Merck Index 15, 7845
Formula Weight 320.51
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material 5β-Pregnane-3α, 20α-diol, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.