accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 8000-73-5

IUPAC Name: diammonium carbamate hydrogen carbonate
Synonyms: diammonium bicarbonate carbamate
Molecular Weight (g/mol): 157.13
Molecular Formula: C2H11N3O5
InChi Key: YNSUIDWLZCFLML-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].NC([O-])=O.OC([O-])=O
Grade 
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (6)
  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)