Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Carbonate, Powder, NF, 30-34%, Spectrum™ Chemical
SDP

Catalog No. 18600578 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-578 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-600-578 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1630500GM

missing translation for 'validMainframeMsg'

A1630, 8000-73-5

Spectrum™ Chemical Ammonium Carbonate, Powder, NF is an excipient used as an alkalinizing agent in liquid preparations for product stability. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8000-73-5
Molecular Formula C2H11N3O5
Molecular Weight (g/mol) 157.13
InChI Key YNSUIDWLZCFLML-UHFFFAOYSA-N
IUPAC Name diammonium carbamate hydrogen carbonate
SMILES [NH4+].[NH4+].NC([O-])=O.OC([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 30 to 34%
Grade NF
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.