CAS RN 8001-35-2
IUPAC Name:
1,4,5,6,7,7-hexachloro-2,2-bis(chloromethyl)-3-methylidenebicyclo[2.2.1]heptane
Synonyms:
toxaphene
Molecular Weight (g/mol):
411.77
Molecular Formula:
C10H8Cl8
InChi Key:
OEJNXTAZZBRGDN-UHFFFAOYNA-N
SMILES:
ClCC1(CCl)C(=C)C2(Cl)C(Cl)C(Cl)C1(Cl)C2(Cl)Cl
1
–
6
of
6
results
1
Toxaphene Solution, 1000μg/mL in methanol, Ultra Scientific
CAS: 8001-35-2 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 MDL Number: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N IUPAC Name: methanol SMILES: CO
4
Toxaphene, SPEX CertiPrep™
CAS: 8001-35-2
5
Toxaphene Standard, High Level, SPEX CertiPrep™
CAS: 8001-35-2
6
Toxaphene, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material