Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toxaphene (For EPA 500, 600, 8000/; Appendix IX, CLP Methods)

Catalog No. USPP2711
Encompass
Change view
Click to view available options
Quantity:
1 mL (in Hexane)
1 mL (in Methanol)
Packaging:
Ampule
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
US-PP-2711 1 mL (in Hexane) Ampule
US-PP-2701 1 mL (in Methanol) Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. US-PP-2711 Supplier Agilent Technologies Supplier No. PP2711
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Toxaphene at 100μg/mL in methanol or hexane

Chemical Identifiers

CAS 8001-35-2

Specifications

Packaging Ampule
Methods For EPA 500, 600, 8000/Appendix IX
Quantity 1 mL (in Hexane)
Chemical Name or Material Toxaphene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.