accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOOOH

CAS RN 8002-74-2

IUPAC Name: 1-{2-[2-hydroxy-3-(propylamino)propoxy]phenyl}-3-phenylpropan-1-one
Synonyms: propafenone (s)-propafenone propafenon hexal propafenone, (R)-isomer propafenona propafenone (substance) (s)-isomer propafenone (R)-isomer propafenone (+-)-propafenone baxarytmon
Molecular Weight (g/mol): 341.45
Molecular Formula: C21H27NO3
InChi Key: JWHAUXFOSRPERK-UHFFFAOYNA-N
SMILES: CCCNCC(O)COC1=CC=CC=C1C(=O)CCC1=CC=CC=C1
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (11)
  • (4)
  • (1)
Grade 
  • (1)
  • (4)
  • (2)
  • (7)
Physical Form 
  • (1)
  • (4)
  • (8)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (11)
  • (1)
  • (1)
  • (5)
  • (11)
  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (11)
  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (7)
  • (1)
  • (4)
  • (8)
  • (1)
  • (2)
  • (1)