Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wax Paraffin, White, Cake, Spectrum™ Chemical
SDP

Catalog No. 18607741 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 lb.
22.7 kg
25 kg
12 x 1 lb.
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-741 25 kg Poly Bottle
18-607-738 1 lb. Poly Bottle
18-607-739 12 x 1 lb. Poly Bottle
18-607-740 22.7 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-741 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. W102525KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

W1025, 8002-74-2

Spectrum™ Chemical Wax Paraffin, White, Cake consists of a mix of hydrocarbon molecules and is derivable from coal, petroleum, or oil shale. It is used in electrical insulation, lubrication and candles. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 8002-74-2
Molecular Formula C21H27NO3
Molecular Weight (g/mol) 341.45
InChI Key JWHAUXFOSRPERK-UHFFFAOYNA-N
IUPAC Name 1-{2-[2-hydroxy-3-(propylamino)propoxy]phenyl}-3-phenylpropan-1-one
SMILES CCCNCC(O)COC1=CC=CC=C1C(=O)CCC1=CC=CC=C1

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 25 kg
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Wax Paraffin, White, Cake
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.