accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 8003-59-6

NaOSOOHNNO(Z)

CAS RN 8003-59-6

IUPAC Name: sodium 6-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}naphthalene-1-sulfonate
Synonyms: sodium 6-{2-[(1Z)-2-oxonaphthalen-1-ylidene]hydrazin-1-yl}naphthalene-1-sulfonate
Molecular Weight (g/mol): 400.38
Molecular Formula: C20H13N2NaO4S
InChi Key: PNQGDQXHSLQMMM-KGYDJYTLSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=C2C=CC(N\N=C3/C(=O)C=CC4=CC=CC=C34)=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results