Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Red 9, TCI America™
SDP

Catalog No. S014025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S014025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S014025G Supplier TCI America Supplier No. S014025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 8003-59-6
Molecular Formula C20H13N2NaO4S
Molecular Weight (g/mol) 400.384
MDL Number MFCD00151319
InChI Key GUJSXTXHVQUPDG-KGYDJYTLSA-M
Synonym Silk Scarlet
PubChem CID 133112856
IUPAC Name sodium;6-[(2Z)-2-(2-oxonaphthalen-1-ylidene)hydrazinyl]naphthalene-2-sulfonate
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC4=C(C=C3)C=C(C=C4)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 400.38
Chemical Name or Material Acid Red 9
EINECSNumber (5)-4293
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.