accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHNH2

CAS RN 80126-54-1

IUPAC Name: 2-amino-3-(2-methylphenyl)propanoic acid
Synonyms: 2-amino-3-(2-methylphenyl)propanoic acid
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: NHBKDLSKDKUGSB-UHFFFAOYNA-N
SMILES: CC1=CC=CC=C1CC(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results