accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 8013-17-0

IUPAC Name: 1,3,4,5,6-pentahydroxyhexan-2-one; 2,3,4,5,6-pentahydroxyhexanal
Synonyms: inverted sugar syrup saccharide hydrolysate
Molecular Weight (g/mol): 360.31
Molecular Formula: C12H24O12
InChi Key: PJVXUVWGSCCGHT-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C(O)C(O)C=O.OCC(O)C(O)C(O)C(=O)CO
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results