accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)OO(R)H3C(S)OH3COH3CHOCH3O(S)H3C(R)OOH3COHNCH3H3C(R)H3COH(R)CH3NOOOCH3(S)H3C(R)OH(S)CH3HO(E)

CAS RN 80214-83-1

IUPAC Name: (3R,4S,5S,6R,7R,9R,10E,11S,12R,13S,14R)-6-{[4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-[(5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl)oxy]-3,5,7,9,11,13-hexamethyl-10-(2,4,7-trioxa-1-azaoctan-1-ylidene)-1-oxacyclotetradecan-2-one
Synonyms: roxithromycin
Molecular Weight (g/mol): 837.06
Molecular Formula: C41H76N2O15
InChi Key: RXZBMPWDPOLZGW-KMAKEOJNSA-N
SMILES: CC[C@H]1OC(=O)[C@H](C)[C@@H](OC2CC(C)(OC)C(O)C(C)O2)[C@H](C)[C@@H](OC2OC(C)CC(C2O)N(C)C)[C@](C)(O)C[C@@H](C)\C(=N/OCOCCOC)[C@H](C)[C@@H](O)[C@]1(C)O
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)