Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roxithromycin, G-Biosciences
SCI_ED

Catalog No. S000021
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000021 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000021 Supplier G-Biosciences Supplier No. RC1095
Add to Cart
Add to Cart

Roxithromycin is a macrolide antibiotic and derivative of erythromycin. It targets both gram-negative and gram-positive bacteria and has shown strong efficacy against Legionella pneumophila and a variety of oral pathogens

Roxithromycin inhibits the activation of NADPH oxidase and promotes neutrophil apoptosis.

Chemical Identifiers

CAS 80214-83-1

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 10 g
Percent Purity >95%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.