CAS RN 8028-48-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Promotions Available
Orange Oil, Florida Type, Coldpressed, FCC, 1.2-2.5%, Spectrum™ Chemical
Orange Oil, Florida Type, Coldpressed, FCC, 1.2-2.5%, Spectrum™ Chemical
CAS: 8028-48-6
2
3
Promotions Available
Orange Oil, California Type, Coldpressed, FCC, 1.2-2.5%, Spectrum™ Chemical
Orange Oil, California Type, Coldpressed, FCC, 1.2-2.5%, Spectrum™ Chemical
CAS: 8028-48-6