CAS RN 80370-57-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Chemicals Ceftiofur
CAS: 80370-57-6 Molecular Formula: C19H17N5O7S3 Molecular Weight (g/mol): 523.55