accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80382-23-6

H2OH2ONaCH3OOO

CAS RN 80382-23-6

IUPAC Name: sodium 2-{4-[(2-oxocyclopentyl)methyl]phenyl}propanoate dihydrate
Synonyms: sodium 2-{4-[(2-oxocyclopentyl)methyl]phenyl}propanoate dihydrate
Molecular Weight (g/mol): 304.32
Molecular Formula: C15H21NaO5
InChi Key: BAZQYVYVKYOAGO-UHFFFAOYNA-M
SMILES: O.O.[Na+].CC(C([O-])=O)C1=CC=C(CC2CCCC2=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results